Learn More
Sino Biological EBOV EBOV-NP MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40444MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (Sudan ebolavirus, strain Gulu) Nucleoprotein / NP ( Catalog#40444-V07E1; YP_138520.1; Gly630-Asp738). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2186
 Restricted Item