Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP Antibody, Rabbit PAb
Supplier:  Sino Biological 40443RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Nucleoprotein / NP ( Catalog#40443-V07E1; AHX24646.1; His630-Gln739). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8700