Learn More
Sino Biological Ebola virus EBOV (Sudan ebolavirus, strain Gulu) Nucleoprotein / NP Antibody, Rabbit PAb
Supplier:  Sino Biological 40444RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (Sudan ebolavirus, strain Gulu) Nucleoprotein / NP ( Catalog#40444-V07E1; YP_138520.1; Gly630-Asp738). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2229