Learn More
Sino Biological EBOV EBOV-VP24 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40456MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 ( Catalog#40456-V07E; YP_003815439.1; Met1-Lys233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1439