Learn More
Sino Biological EBOV EBOV-VP24 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 40454T4620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 ( Catalog#40454-V07E; AHX24653.1; Met1-Gly233). Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 specific IgG was purified by Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 affinity chromatography.
Catalog No. 50-237-2197