Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 Antibody, Rabbit PAb
Supplier:  Sino Biological 40454RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) VP24 (Catalog#40454-V07E; AHX24653.1; Met1-Gly233). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2163