Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40456T4620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 ( Catalog#40456-V07E; YP_003815439.1; Met1-Lys233). Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 specific IgG was purified by Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 affinity chromatography.
Catalog No. 50-241-2232