Learn More
Sino Biological EBOV EBOV-VP40 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40446MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Matrix protein VP40 ( Catalog#40446-V07E; AHX24648.1; Asn31-Lys326). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1376