Learn More
Sino Biological EBOV EBOV-VP40 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40448MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40 / Matrix protein VP40 ( Catalog#40448-V07E; YP_003815434.1; Ala31-Lys326). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2256