Learn More
Sino Biological Ebola virus EBOV (subtype Sudan, strain Gulu) VP40 / Matrix protein VP40 Antibody, Rabbit PAb
Supplier:  Sino Biological 40447RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Sudan, strain Gulu) VP40 / Matrix protein VP40 (Catalog#40447-V07E; YP_138522.1; Val31-Lys326). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2526