Learn More
Sino Biological Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Matrix protein VP40 Antibody, Rabbit PAb
Supplier:  Sino Biological 40446RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant EBOV Ebola virus EBOV (subtype Zaire, strain H.sapiens-wt/GIN/2014/Kissidougou-C15) Matrix protein VP40 (Catalog#40446-V07E; AHX24648.1; Asn31-Lys326). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1933