Learn More
Sino Biological Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40 / Matrix protein VP40 Antibody, Rabbit PAb
Supplier:  Sino Biological 40448RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40 / Matrix protein VP40 ( Catalog#40448-V07E; YP_003815434.1; Ala31-Lys326). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2315