Learn More
Sino Biological Ebola virus EBOV (subtype Sudan, strain Gulu) VP40 / Matrix protein VP40 Antibody, Mouse MAb
Supplier:  Sino Biological 40447MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Sudan, strain Gulu) VP40 / Matrix protein VP40 ( Catalog#40447-V07E; YP_138522.1; Val31-Lys326). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-3717