Learn More
Aviva Systems Biology Corp EBP Antibody - N-terminal region (ARP46742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EBP. Alternate names: CDPX2, CHO2, CPX, CPXDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53488