Learn More
Enzo Life Sciences EBP50 (human) monoclonal antibody (EBP-10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804438C100
Encompass_Preferred
Clone: EBP-10. Isotype: IgG2b. Immunogen: Recombinant full length human EBP50 (ERM (ezrin/radixin/moesin)-binding phosphoprotein of 50kDa). Species reactivity: Human. Applications: IHC (PS), IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-200-9910