Learn More
Electron Microscopy Sciences EBSD Specimen Holder, 25 mm
SDP
Supplier:  Electron Microscopy Sciences 7594411
Encompass
Made from aluminum, this holder for EBSD has a built-in 70 degree pre-tilt angle . Comes complete with stainless steel screws and allen key. Accommodates 1" (25mm) mounts.
Size: 25 mm
Catalog No. 50-192-8200