Learn More
Electron Microscopy Sciences EBSD Specimen Holder, M4 Compatible, 25 mm
SDP
Catalog No. 501928201
Encompass
Made from aluminum, this holder for EBSD has a built-in 70 degree pre-tilt angle . Comes complete with stainless steel screws and allen key. Accommodates 1" (25mm) mounts.
Size: 25 mm
Catalog No. 50-192-8201 Supplier Electron Microscopy Sciences Supplier No. 7594412
Made from aluminum, this holder for EBSD has a built-in 70 degree pre-tilt angle . Comes complete with stainless steel screws and allen key. Accommodates 1" (25mm) mounts.
Size: 25 mm