Learn More
Electron Microscopy Sciences EBSD Specimen Holder, M4 Compatible, 32 mm
SDP
Catalog No. 501928203
Encompass
Made from aluminum, this holder for EBSD has a built-in 70 degree pre-tilt angle . Comes complete with stainless steel screws and allen key. Accommodates 1.25" (32mm) mounts.
Size: 32 mm
Catalog No. 50-192-8203 Supplier Electron Microscopy Sciences Supplier No. 7594414
Made from aluminum, this holder for EBSD has a built-in 70 degree pre-tilt angle . Comes complete with stainless steel screws and allen key. Accommodates 1.25" (32mm) mounts.
Size: 32 mm