Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ EBSS, calcium, magnesium, phenol red
GREENER_CHOICE

Catalog No. 24010043
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-010-043 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 24-010-043 Supplier Gibco™ Supplier No. 24010043
  • $27.75
    $25.65 / Each of 1

    Save $2.10

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation.

This EBSS is modified as follows:

With: Glucose, Sodium Bicarbonate, Calcium, Magnesium, Phenol Red

The complete formulation is available.

Product Intended Use

For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type Earle's Balanced Salt Solution (EBSS)
Sterility Sterile-filtered
With Additives Phenol Red, Glucose, Sodium Bicarbonate, Calcium, Magnesium
Without Additives No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Osmolality 270 - 310 mOsm/kg
pH 7 to 7.4
Content And Storage Storage conditions: 15-30° C.
Shipping conditions: Ambient.
Shelf life: 36 months from date of manufacture.
Show More Show Less
Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Is it true that phenol red has estrogenic properties?

Phenol red in tissue culture is a weak estrogen. Implications concerning study of estrogen-responsive cells in culture are discussed in Proc Natl Acad Sci U S A 83:2496 (1986).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.