Learn More
Medchemexpress LLC Ebsulfur | 2527-03-9 | 99.4% | 1 ML

Supplier: Medchemexpress LLC HY12817010MM/1ML
Ebsulfur is a benzisothiazolone cytocidal inhibitor. It targets trypanothione reductase (TryR) in *Trypanosoma brucei*, with IC50 values ranging from 61-293 nM for this organism. This compound is used in the research of African trypanosomiasis.
- Targets trypanothione reductase (TryR) of *Trypanosoma brucei*
- Used for research of African trypanosomiasis
- Soluble in DMSO at 62.5 mg/mL
- Solid, white to off-white appearance
- Purity of 99.4%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.