Learn More
Medchemexpress LLC Ebsulfur | 2527-03-9 | 99.4% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128170
Encompass_Preferred
Ebsulfur is a benzisothiazolone cytocidal inhibitor targeting the trypanothione reductase (TryR) of Trypanosoma brucei, with IC50 values of 61-293 nM, making it suitable for African trypanosomiasis research. It is a white to off-white solid with a molecular weight of 227.28 and formula C13H9NOS. For in vitro use, it has a solubility of 62.5 mg/mL (274.99 mM) in DMSO, requiring ultrasonic dissolution with newly opened DMSO due to its hygroscopic nature.
  • Potent cytocidal inhibitor of trypanothione reductase (TryR) in Trypanosoma brucei.
  • Useful for research on African trypanosomiasis.
  • High purity of 99.43%.
  • Available in various solid quantities and as a 10 mM solution in DMSO.
  • Includes detailed documentation (data sheet, COA, SDS, HNMR, LCMS, handling instructions).
Catalog No. 50-003-42701