Learn More
Medchemexpress LLC Ebsulfur | 2527-03-9 | 99.4% | 50 MG
SDP
Catalog No. 5000339197
Encompass_Preferred
Ebsulfur is a benzisothiazolone cytocidal inhibitor that targets the trypanothione reductase (TryR) of *Trypanosoma brucei*. It is used for research on African trypanosomiasis.
  • Targets trypanothione reductase (TryR) of *Trypanosoma brucei*
  • Has IC50 values of 61-293 nM for *Trypanosoma brucei*
  • Used for research on African trypanosomiasis
Catalog No. 50-003-39197 Supplier Medchemexpress LLC Supplier No. HY12817050MG
Ebsulfur is a benzisothiazolone cytocidal inhibitor that targets the trypanothione reductase (TryR) of *Trypanosoma brucei*. It is used for research on African trypanosomiasis.
  • Targets trypanothione reductase (TryR) of *Trypanosoma brucei*
  • Has IC50 values of 61-293 nM for *Trypanosoma brucei*
  • Used for research on African trypanosomiasis