Learn More
Cayman Chemical (−)-Eburnamonine, Purity: ≥98%, CAS: 4880-88-0, 50 mg
SDP
Catalog No. 502916102
Encompass_Preferred
An indole alkaloid with vasodilator and neuroprotective activities; increases cerebral blood flow and capillary circulation in anesthetized dogs at 2.5 mg/kg; prevents scopolamine-induced amnesia in a passive avoidance test in mice at 20 mg/kg; enhances acquisition of the conditioned avoidance response induced by hypobaric hypoxia in rats at 20 mg/kg; increases the onset time to hypoxia-induced brain death in rats at 0.25 mg/kg per minute; increases survival time and inhibits decreases in body temperature induced by hypobaric hypoxia in rats at 45 mg/kg
Catalog No. 50-291-6102 Supplier Cayman Chemical Supplier No. 3756950 MG
An indole alkaloid with vasodilator and neuroprotective activities; increases cerebral blood flow and capillary circulation in anesthetized dogs at 2.5 mg/kg; prevents scopolamine-induced amnesia in a passive avoidance test in mice at 20 mg/kg; enhances acquisition of the conditioned avoidance response induced by hypobaric hypoxia in rats at 20 mg/kg; increases the onset time to hypoxia-induced brain death in rats at 0.25 mg/kg per minute; increases survival time and inhibits decreases in body temperature induced by hypobaric hypoxia in rats at 45 mg/kg