Learn More
Enzo Life Sciences (-)-Eburnamonine (100mg). CAS: 4880-88-0
SDP
Supplier:  Enzo Life Sciences ALX350216M100
Encompass_Preferred
Cerebral metabolic stimulant; shows antihypoxic effects. Vasodilator. Alternative name: (3alpha,16alpha)-Eburnamenin-14(15H)-one, Vincamone, Vinburnine. Purity: ≥99% (Assay). Solubility: Soluble in chloroform. Long Term Storage: -20°C.
Catalog No. 50-200-8781