Learn More
Abnova™ EBV CISH Probe
Catalog No. 89335575 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-575 CISH-P
1 options
Catalog No. 89-335-575 Supplier Abnova™ Supplier No. CG0024
Add to Cart
Add to Cart
EBV CISH Probe is designed for the qualitative detection of Epstein-Barr virus (EBV) EBER RNA in formalin-fixed, paraffin-embedded specimens, such as lymphomas, by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation HRP-DAB Kit, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
A positive reactivity for Epstein-Barr virus (EBV) EBER RNA in the target cells is indicated by a distinctly stained nucleus. Visualization of signals should be performed at least at 100-fold magnification resulting in easily visible signals. Do not evaluate areas of necrosis, overlapping nuclei, over-digested nuclei, and nuclei with weak signal intensity.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (55°C) and mix briefly before use.
For Use With (Application) CISH-P
Includes Reagent Provided:
Digoxigenin-labeled oligonucleotides targeting mRNA sequences encoding EBER-1 and EBER-2 regions.