Learn More
Drg International Inc DRG EBV-EBNA-1 IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA4246R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Epstein-Barr virus nuclear antigen type 1 (EBV-EBNA-1) in serum. This product is for Research Use Only. The DRG Epstein-Barr Virus (EBNA-1) IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with recombinant EBV nuclear antigen type 1. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7608