Learn More
Sino Biological EBV EBV-gp350 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 40373RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Epstein-Barr virus (Herpesvirus 4) EBV Glycoprotein gp350 / EBV GP350 (Catalog#40373-V08H; YP_401667.1; Met1-Pro490). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2931