Learn More
Sino Biological Epstein-Barr virus (strain B95-8) EBV Lytic switch protein BZLF1 Protein (His & MBP Tag)
Catalog No. 502993302
Encompass_Preferred
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) EBV Lytic switch protein BZLF11 (P03206) (Met1-Phe245) was expressed with a N-terminal polyhistidine tag followed by a MBP tag.
Catalog No. 50-299-3302 Supplier Sino Biological Supplier No. 41059V10E100
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) EBV Lytic switch protein BZLF11 (P03206) (Met1-Phe245) was expressed with a N-terminal polyhistidine tag followed by a MBP tag.