Learn More
Macherey-Nagel HPLC column EC 125/2 NUCLEODUR 100-3 C8 ec
Supplier:  Macherey-Nagel 760060.20
Encompass_Preferred
EC HPLC column - EC 125/2 NUCLEODUR 100-3 C8 ec - length: 125 mm, ID: 2 mm - pack of 1
Catalog No. NC1540417