Learn More
Macherey-Nagel HPLC column EC 125/4.6 NUCLEOSIL 100-7 C18
Supplier:  Macherey-Nagel 720951.46
Encompass_Preferred
EC HPLC column - EC 125/4.6 NUCLEOSIL 100-7 C18 - length: 125 mm, ID: 4.6 mm - pack of 1
Catalog No. NC0726679