Learn More
Medchemexpress LLC EC-17 disodium salt | 910661-33-5 | 96.3% | 916.82 | 25 MG
SDP
Catalog No. 5000351584
Encompass_Preferred
EC-17 disodium salt is a folate receptor alpha (FRα) targeting contrast agent that exhibits fluorescent properties within the visible light spectrum. Its maximum excitation and emission wavelengths are 490 nm and 520 nm, respectively. It is intended for research use only.
  • Folate receptor alpha (FRα) targeting contrast agent
  • Fluorescent properties in the visible light spectrum
  • Maximum excitation wavelength: 490 nm
  • Maximum emission wavelength: 520 nm
Catalog No. 50-003-51584 Supplier Medchemexpress LLC Supplier No. 25MGHY13615A
EC-17 disodium salt is a folate receptor alpha (FRα) targeting contrast agent that exhibits fluorescent properties within the visible light spectrum. Its maximum excitation and emission wavelengths are 490 nm and 520 nm, respectively. It is intended for research use only.
  • Folate receptor alpha (FRα) targeting contrast agent
  • Fluorescent properties in the visible light spectrum
  • Maximum excitation wavelength: 490 nm
  • Maximum emission wavelength: 520 nm