Learn More
Macherey-Nagel HPLC column EC 250/4.6 NUCLEOSIL 50-5
Supplier:  Macherey-Nagel 720093.46
Encompass_Preferred
EC HPLC column - EC 250/4.6 NUCLEOSIL 50-5 - length: 250 mm, ID: 4.6 mm - pack of 1
Catalog No. NC1513828