Learn More
Macherey-Nagel HPLC guard column EC 4/3 NUCLEOSIL 100-5 C18
Catalog No. NC1186319
Encompass_Preferred
HPLC guard column - EC 4/3 NUCLEOSIL 100-5 C18 - length: 4 mm, ID: 3 mm - pack of 3 - require Column Protection System guard column holder (REF 718966) - for analytical columns with 3, 4 and 4.6 mm ID
Catalog No. NC1186319 Supplier Macherey-Nagel Supplier No. 721074.30
HPLC guard column - EC 4/3 NUCLEOSIL 100-5 C18 - length: 4 mm, ID: 3 mm - pack of 3 - require Column Protection System guard column holder (REF 718966) - for analytical columns with 3, 4 and 4.6 mm ID