Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ EC Bottled Broth with Durham tubes 9mL Vials

Catalog No. 10486164 Shop All GVS Products
Change view
Click to view available options
20pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
10-486-164 20pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-486-164 Supplier GVS™ Supplier No. 10496714
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EC (Escherichia coli) Broth is used to detect Escherichia coli (E-Coli) and coliforms.

Description

  • EC Broth contains casein peptone as a source of nutrients. Lactose provides the carbohydrate fermented by coliform bacteria
    and Escherichia coli (E.Coli). In addition, lactose-positive bacteria metabolize lactose with gas formation. Gram-positive bacteria are inhibited by the mixture of bile salts.

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 9 mL Portions, Reduce Risk of Cross-Contamination.
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • for the detection of Escherichia coli in water and food samples by fluorogenic means.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits.
This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Appearance Clear Yellow
ATCC® 25922
Broth Type EC Bottled Broth
Cap Color Black
Cap Type Phenolic with Rubber Liner
Certifications/Compliance Batch-specific quality certificate in each pack
Color Clear Yellow
Content And Storage 2°C to 8°C
Country of Origin United States
Detects coliforms and E.coli.
Form Liquid
Format Vial
Manufacturing Origin United States
Media Type EC Broth
Product Type Broth
Sterility Sterile
Target Organism Class Coliforms
Test Type Water Monitoring
Fill Volume 9.0 mL
Final Product Type Liquid Medium
Formulation includes casein peptone, lactose, and bile salts.
For Use With (Application) Detection of Fecal Coliforms in Water and Wastewater
Includes Durham Tubes
Incubation Conditions 44.5°C ±2°C
Incubation Time 24 hr.
Material Borosilicate Glass
No. per Pack 20
Packaging Type Vial
Quantity 9 mL
Shipping Condition 2°C to 8°C
Specimen Type Water, Wastewater
Height (Metric) 65 mm
Diameter (Metric) 19 mm
Protein Source Casein
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.