Learn More
Bioanalytical Systems Inc EC C-3 CELL LOW VLM CELL KT
SDP
Catalog No. 50854563
Encompass_Preferred
Kit, C3 Low Volume Cell; Includes: cell vial, cell top, sample chamber and o-ring, reference electrode, auxiliary electrode, (2) port plug, and replacement vycor frits *Does not ship to Puerto Rico
Catalog No. 50-854-563 Supplier Bioanalytical Systems Inc Supplier No. MF2041
Kit, C3 Low Volume Cell; Includes: cell vial, cell top, sample chamber and o-ring, reference electrode, auxiliary electrode, (2) port plug, and replacement vycor frits *Does not ship to Puerto Rico