Learn More
Bioanalytical Systems Inc EC C3 CELL STAND
SDP
Catalog No. 50854249
Encompass
Stand, C-3 Cell; BASi; General purpose accessory for electroanalytical experiements; Holds electrochemical cell components; Allows stirring/purging of analyte solution; Manual or remote control *Does not ship to Puerto Rico
Catalog No. 50-854-249 Supplier Bioanalytical Systems Inc Supplier No. EF1085
Add to Cart
Add to Cart
Request a Quote
Stand, C-3 Cell; BASi; General purpose accessory for electroanalytical experiements; Holds electrochemical cell components; Allows stirring/purging of analyte solution; Manual or remote control *Does not ship to Puerto Rico