Learn More
Bioanalytical Systems Inc EC CELL CON 0.040 MALE
SDP
Catalog No. 50854540
Encompass_Preferred
Connectors, Cable; For EC Products; 0.040 in. male cell connector; Gold-plated brass; For flow cell working electrodes; 12/pk *Does not ship to Puerto Rico
Catalog No. 50-854-540 Supplier Bioanalytical Systems Inc Supplier No. MF2002
Connectors, Cable; For EC Products; 0.040 in. male cell connector; Gold-plated brass; For flow cell working electrodes; 12/pk *Does not ship to Puerto Rico