Learn More
Bioanalytical Systems Inc EC CGME-MERC COLLECTOR ROD
SDP
Catalog No. 50854584
Encompass_Preferred
Electrode; Mercury Drop; Mercury Collector Rod; For spill management; For Use With: BASi Controlled Growth Mercury Electrode; Contains mercury *Does not ship to Puerto Rico
Catalog No. 50-854-584 Supplier Bioanalytical Systems Inc Supplier No. MF2069
Electrode; Mercury Drop; Mercury Collector Rod; For spill management; For Use With: BASi Controlled Growth Mercury Electrode; Contains mercury *Does not ship to Puerto Rico