Learn More
Bioanalytical Systems Inc EC COILED PT WIRE AUX ELECT
SDP
Catalog No. 50854893
Encompass_Preferred
Electrode, Platinum Wire Auxiliary; Coiled; 0.5mm dia. x 23cm L; With gold-plated connector; Mounted in CTFE cylinder; For use with RDE-1, RDE-2 and bulk electrolysis cell *Does not ship to Puerto Rico
Catalog No. 50-854-893 Supplier Bioanalytical Systems Inc Supplier No. MW1033
Electrode, Platinum Wire Auxiliary; Coiled; 0.5mm dia. x 23cm L; With gold-plated connector; Mounted in CTFE cylinder; For use with RDE-1, RDE-2 and bulk electrolysis cell *Does not ship to Puerto Rico