Learn More
Bioanalytical Systems Inc EC ELECT-AGE VOLTAMMETRY
SDP
Catalog No. 50854546
Encompass_Preferred
Electrode, Stationary; Voltammetry; Silver (AGE); 1.6mm dia. *Does not ship to Puerto Rico
Catalog No. 50-854-546 Supplier Bioanalytical Systems Inc Supplier No. MF2011
Electrode, Stationary; Voltammetry; Silver (AGE); 1.6mm dia. *Does not ship to Puerto Rico