Learn More
Bioanalytical Systems Inc EC ELECT-CPE-VOLTAMMETRY
SDP
Catalog No. 50854545
Encompass_Preferred
Electrode, Body; Stationary; Voltammetry; For 3.0mm ID Carbon Paste Electrode (CPE) *Does not ship to Puerto Rico
Catalog No. 50-854-545 Supplier Bioanalytical Systems Inc Supplier No. MF2010
Electrode, Body; Stationary; Voltammetry; For 3.0mm ID Carbon Paste Electrode (CPE) *Does not ship to Puerto Rico