Learn More
Bioworld EC Broth Medium, 500 g
Catalog No. 50254425
Encompass_Preferred
EC MediumA selective broth medium used to detect the presence of coliform bacteria at 37 °C and Escherichia coli at 44.5 °CPrepared Appearance: light amber brothSoluble in warm water37g/LComposition:Pancreatic Digest of Casein Bile Salts Mixture Lactose Dipotassium Phosphate Monopotassium Phospate Sodium Chloride
Catalog No. 50-254-425 Supplier Bioworld Supplier No. 306210031
EC MediumA selective broth medium used to detect the presence of coliform bacteria at 37 °C and Escherichia coli at 44.5 °CPrepared Appearance: light amber brothSoluble in warm water37g/LComposition:Pancreatic Digest of Casein Bile Salts Mixture Lactose Dipotassium Phosphate Monopotassium Phospate Sodium Chloride