Learn More
Hach Company EC Medium MPN Tubes, pk/15
Catalog No. 501994928 Shop All Hach Company Products
Encompass
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
Catalog No. 50-199-4928 Supplier Hach Company Supplier No. 1410415
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.