Learn More
Bioworld EC Medium w/ MUG, 500 g
Catalog No. 50254426
Encompass_Preferred
EC MediumA selective broth medium used to detect the presence of Escherichia coli in water and food samples by fluorogenic procedurepH: 6.9Dehydrated Appearance: light beige, homogeneous powder Prepared Appearance: light amber brothSoluble in waterComposition:Pancreatic Digest of Casein Lactose Bile Salt Mixture Dipotassium Phosphate Monopotassium Phosphate Sodium Chloride MUG Reagent
Catalog No. 50-254-426 Supplier Bioworld Supplier No. 306210041
EC MediumA selective broth medium used to detect the presence of Escherichia coli in water and food samples by fluorogenic procedurepH: 6.9Dehydrated Appearance: light beige, homogeneous powder Prepared Appearance: light amber brothSoluble in waterComposition:Pancreatic Digest of Casein Lactose Bile Salt Mixture Dipotassium Phosphate Monopotassium Phosphate Sodium Chloride MUG Reagent