Learn More
Bioanalytical Systems Inc 50818663QUS REF ELEC KI-AG/AG
SDP
Catalog No. 50854578
Encompass_Preferred
Kit, Non-aqueous reference electrode; Bioanalytical; Glass electrode body; Porous Vycor tip attached w/heat shrink tubing; Removable PTFE top w/silver wire, 0.060 in. gold-plated connecting pin; Solid silver nitrate, heat shrink tubing *Does not ship to Puerto Rico
Catalog No. 50-854-578 Supplier Bioanalytical Systems Inc Supplier No. MF2062
Kit, Non-aqueous reference electrode; Bioanalytical; Glass electrode body; Porous Vycor tip attached w/heat shrink tubing; Removable PTFE top w/silver wire, 0.060 in. gold-plated connecting pin; Solid silver nitrate, heat shrink tubing *Does not ship to Puerto Rico