Learn More
Bioanalytical Systems Inc EC PT WIRE AUXILLARY 6CM
SDP
Catalog No. 50854911
Encompass_Preferred
Electrode, Platinum Wire Auxiliary; 0.5mm dia. x 6cm L; With gold-plated connector; Mounted in CTFE cylinder; For C-1 and C-2 cell stands and VC-2 voltammetry cell *Does not ship to Puerto Rico
Catalog No. 50-854-911 Supplier Bioanalytical Systems Inc Supplier No. MW4130
Electrode, Platinum Wire Auxiliary; 0.5mm dia. x 6cm L; With gold-plated connector; Mounted in CTFE cylinder; For C-1 and C-2 cell stands and VC-2 voltammetry cell *Does not ship to Puerto Rico