Learn More
Bioanalytical Systems Inc EC RDE-2 ROTATING ELECTRODE
SDP
Catalog No. 50854250
Encompass
Electrode, Rotating Disk; BASi RDE-2; 1/4 in. dia.; Rotation from 50 to 10,000 RPM; Acc.+1 percent; Sm. sample vol.; Standard addition port *Does not ship to Puerto Rico
Catalog No. 50-854-250 Supplier Bioanalytical Systems Inc Supplier No. EF1100
Add to Cart
Add to Cart
Request a Quote
Electrode, Rotating Disk; BASi RDE-2; 1/4 in. dia.; Rotation from 50 to 10,000 RPM; Acc.+1 percent; Sm. sample vol.; Standard addition port *Does not ship to Puerto Rico