Learn More
MOLTOX Molecular Toxicology Inc EC TRI PC, 5 plates/sleeve, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 21199
Encompass
For phenotype confirmation of E. coli WP2. Tests for tryptophan auxotrophy & pKM101 presence in the Ames assay. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs. If your laboratory uses a specifically modified formulation or packaging, contact us for a full assessment.
Catalog No. 50-948-608