Learn More
Bioanalytical Systems Inc EC U-ELECT-MAUE AU-10UM DIA
SDP
Catalog No. 50854543
Encompass_Preferred
Microelectrode; MAUE gold; 10um diameter; For use with Low Current Module *Does not ship to Puerto Rico
Catalog No. 50-854-543 Supplier Bioanalytical Systems Inc Supplier No. MF2006
Microelectrode; MAUE gold; 10um diameter; For use with Low Current Module *Does not ship to Puerto Rico