Learn More
Bioanalytical Systems Inc EC VIAL PTFE VOLT CELL
SDP
Catalog No. 50854736
Encompass_Preferred
Vial, Voltammetry Cell; PTFE; Water jacketed; Low volume; 51 x 51mm; 2-15mL; For C-3 Cell Stand, CGME, and RDE-2 *Does not ship to Puerto Rico
Catalog No. 50-854-736 Supplier Bioanalytical Systems Inc Supplier No. MR1205
Vial, Voltammetry Cell; PTFE; Water jacketed; Low volume; 51 x 51mm; 2-15mL; For C-3 Cell Stand, CGME, and RDE-2 *Does not ship to Puerto Rico